4-(Trifluoromethoxy)benzaldehyde, 96%
4-(Trifluoromethoxy)benzaldehyde, 96%
4-(Trifluoromethoxy)benzaldehyde, 96%
4-(Trifluoromethoxy)benzaldehyde, 96%
Thermo Scientific Chemicals
4-(Trifluoromethoxy)benzaldehyde, 96%
CAS: 659-28-9 | C8H5F3O2 | 190.121 g/mol
Have Questions?
Catalog NumberQuantity
ALFA10671.065 g
Catalog number ALFA10671.06
Price (MYR)
438.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
438.00
EA
Add to cart
Specifications
Chemical Name or Material4- (Trifluoromethoxy)benzaldehyde
CAS659-28-9
Health Hazard 1H227-H302-H315-H319
Health Hazard 2GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3P210-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P370+P378q-P501c