alpha-Benzoin oxime, 98+%
alpha-Benzoin oxime, 98+%
alpha-Benzoin oxime, 98+%
alpha-Benzoin oxime, 98+%
Thermo Scientific Chemicals
alpha-Benzoin oxime, 98+%
CAS: 441-38-3 | C14H13NO2 | 227.26 g/mol
Have Questions?
Catalog NumberQuantity
ALFA10630.1425 g
Catalog number ALFA10630.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Materialalpha-Benzoin oxime
CAS441-38-3
Health Hazard 1H319
Health Hazard 3P264b-P280i-P305+P351+P338
Melting Point151°C to 154°C