1-Bromododecane, 98%
1-Bromododecane, 98%
1-Bromododecane, 98%
1-Bromododecane, 98%
Thermo Scientific Chemicals
1-Bromododecane, 98%
CAS: 143-15-7 | C12H25Br | 249.236 g/mol
Have Questions?
Catalog NumberQuantity
ALFA10600.30250 g
Catalog number ALFA10600.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Bromododecane
CAS143-15-7
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Melting Point-10°C
View more
1-Bromododecane is used as a solvent. It is used as an intermediate to manufacture surfactant, pharmaceuticals and other organic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Bromododecane is used as a solvent. It is used as an intermediate to manufacture surfactant, pharmaceuticals and other organic compounds.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
Giles R. Cokelet; Frederick J. Hollander; Joseph H. Smith. Density and viscosity of mixtures of 1,1,2,2-tetrabromoethane and 1-bromododecane. J. Chem. Eng. Data. 1969, 14, (4) 470-473.