1-Bromononane, 99%
1-Bromononane, 99%
1-Bromononane, 99%
Thermo Scientific Chemicals
1-Bromononane, 99%
CAS: 693-58-3 | C9H19Br | 207.155 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFA10577.22
Price (MYR)
832.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
832.00
EA
Add to cart
Specifications
Chemical Name or Material1-Bromononane
CAS693-58-3
Health Hazard 1H227-H315-H319-H332-H335
Health Hazard 2GHS H Statement
H227
Combustible liquid.
Health Hazard 3P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
View more
1-Bromononane is used as organic intermediates. It is mainly used in pharmaceutical synthesis and industrial dye intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Bromononane is used as organic intermediates. It is mainly used in pharmaceutical synthesis and industrial dye intermediates.
Solubility
Insoluble in water. Soluble in ethyl ether and benzene.
Notes
Stable under normal conditions. Incompatible with oxidizing agents.
RUO – Research Use Only