2,4-Dichloroaniline, 99%
2,4-Dichloroaniline, 99%
2,4-Dichloroaniline, 99%
2,4-Dichloroaniline, 99%
Thermo Scientific Chemicals
2,4-Dichloroaniline, 99%
CAS: 554-00-7 | C6H5Cl2N | 162.013 g/mol
Have Questions?
Catalog NumberQuantity
ALFA10518.22100 g
Catalog number ALFA10518.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material2,4-Dichloroaniline
CAS554-00-7
Health Hazard 1H301+H311+H331-H373-H500
Health Hazard 2GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c