1,2-Naphthoquinone-4-sulfonic acid sodium salt, 97%
1,2-Naphthoquinone-4-sulfonic acid sodium salt, 97%
1,2-Naphthoquinone-4-sulfonic acid sodium salt, 97%
1,2-Naphthoquinone-4-sulfonic acid sodium salt, 97%
Thermo Scientific Chemicals
1,2-Naphthoquinone-4-sulfonic acid sodium salt, 97%
CAS: 521-24-4 | C10H5NaO5S | 260.195 g/mol
Have Questions?
Catalog NumberQuantity
ALFA10507.1425 g
Catalog number ALFA10507.14
Price (MYR)
690.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
690.00
EA
Add to cart
Specifications
Chemical Name or Material1,2-Naphthoquinone-4-sulfonic acid sodium salt
CAS521-24-4
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Colorimetric determination of amino acids and amines1,2-Naphthoquinone-4-sulfonic acid sodium salt is used for the colorimetric determination of amino acids, amines and procaine hydrochloride in pharmaceutical preparations. It is also used in the characterization of phanerochaete chrysosporium. Further, it acts as an intermediate in the synthesis of carbohydrate based napthoquinones, which acts as anticancer agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Colorimetric determination of amino acids and amines1,2-Naphthoquinone-4-sulfonic acid sodium salt is used for the colorimetric determination of amino acids, amines and procaine hydrochloride in pharmaceutical preparations. It is also used in the characterization of phanerochaete chrysosporium. Further, it acts as an intermediate in the synthesis of carbohydrate based napthoquinones, which acts as anticancer agents.
Solubility
Soluble in water and acetone. Slightly soluble in alcohol and ethanol. Insoluble in chloroform.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Muszalska, I.; Sobczak, A.; Kiaszewicz, I.; Rabiega, K.; Lesniewska, M. A.; Jelińska, A. 1,2-Naphthoquinone-4-sulfonic acid sodium salt as a reagent for spectrophotometric determination of rimantadine and memantine. J. Anal. Chem. 2015, 70 (3), 320-327.
  2. Kilan, K.; Warszyński, P. Thickness and permeability of multilayers containing alginate cross-linked by calcium ions. Electrochim. Acta 2014, 144, 254-262.