Diphenylsilanediol, 98%
Diphenylsilanediol, 98%
Diphenylsilanediol, 98%
Diphenylsilanediol, 98%
Thermo Scientific Chemicals
Diphenylsilanediol, 98%
CAS: 947-42-2 | C12H12O2Si | 216.311 g/mol
Have Questions?
Catalog NumberQuantity
ALFA10492.22100 g
Catalog number ALFA10492.22
Price (MYR)
597.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
597.00
EA
Add to cart
Specifications
Chemical Name or MaterialDiphenylsilanediol
Melting Point∼150°C (decomposition)
CAS947-42-2
Health Hazard 1H228-H315-H319-H335
Health Hazard 2GHS H Statement
H228
Flammable solid.
View more
Diphenylsilanediol is used in Medicine field, electronics industry and polymer material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diphenylsilanediol is used in Medicine field, electronics industry and polymer material.
Solubility
Reacts with water.
Notes
Stable under normal temperatures and pressures. Incompatible with Oxidizing agents. Store at room temperature. Air & Light Sensitive.
RUO – Research Use Only
J. Keith Fawcett, Norman Camerman, Arthur Camerman. Stereochemical basis of anticonvulsant drug action. VI. Crystal and molecular structure of diphenylsilanediol. Canadian Journal of Chemistry.1977, 55, (20), 3631-3635.