3-(Trifluoromethyl)benzaldehyde, 97%
3-(Trifluoromethyl)benzaldehyde, 97%
3-(Trifluoromethyl)benzaldehyde, 97%
3-(Trifluoromethyl)benzaldehyde, 97%
Thermo Scientific Chemicals
3-(Trifluoromethyl)benzaldehyde, 97%
CAS: 454-89-7 | C8H5F3O | 174.122 g/mol
Have Questions?
Catalog NumberQuantity
ALFA10463.065 g
Catalog number ALFA10463.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material3- (Trifluoromethyl)benzaldehyde
CAS454-89-7
Health Hazard 1H227-H315-H319-H335
Health Hazard 2GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c