2-Amino-6-methylpyridine, 99%
2-Amino-6-methylpyridine, 99%
2-Amino-6-methylpyridine, 99%
2-Amino-6-methylpyridine, 99%
2-Amino-6-methylpyridine, 99%
CAS: 1824-81-3 | C6H8N2 | 108.14 g/mol
Have Questions?
Catalog NumberQuantity
ALFA10444.22100 g
Catalog number ALFA10444.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Amino-6-methylpyridine
CAS1824-81-3
Health Hazard 1H301-H310-H315-H319-H335
Health Hazard 2GHS H Statement
H301-H310-H315-H319-H335
Toxic if swallowed.
Fatal in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P262-P264b-P270-P271-P280-P301+P310-P302+P350-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P361-P363-P501c