alpha-Methylcinnamaldehyde, predominantly (E), 97%
alpha-Methylcinnamaldehyde, predominantly (E), 97%
alpha-Methylcinnamaldehyde, predominantly (E), 97%
alpha-Methylcinnamaldehyde, predominantly (E), 97%
Thermo Scientific Chemicals
alpha-Methylcinnamaldehyde, predominantly (E), 97%
CAS: 101-39-3 | C10H10O | 146.19 g/mol
Have Questions?
Catalog NumberQuantity
ALFA10429.22100 g
Catalog number ALFA10429.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Materialalpha-Methylcinnamaldehyde, predominantly ( E)
CAS101-39-3
Health Hazard 1H227-H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c