9-Bromofluorene, 97+%
9-Bromofluorene, 97+%
9-Bromofluorene, 97+%
9-Bromofluorene, 97+%
Thermo Scientific Chemicals
9-Bromofluorene, 97+%
CAS: 1940-57-4 | C13H9Br | 245.119 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
5 g
Catalog number ALFA10428.18
Price (MYR)
1,134.00
EA
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
1,134.00
EA
Add to cart
Specifications
Chemical Name or Material9-Bromofluorene
CAS1940-57-4
Health Hazard 1H314-H317
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c
View more
9-Bromofluorene, is used to treat skin dermatological conditions and in pharmaceutical industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
9-Bromofluorene, is used to treat skin dermatological conditions and in pharmaceutical industry.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents. Air sensitive. Stable under normal conditions. Preserve in light-resistant and well-closed containers
RUO – Research Use Only
General References:
  1. CP Defeo. Erythema multiforme bullosum caused by 9-bromofluorene. Arch Dermatol.196694,(5) , 545-551.
  2. G Nie.; T Cai.; L Qu.; J Xu.; Q Wei.; S Zhang. A novel polyfluorene derivative electrodeposited by direct anodic oxidation of 9-bromofluorene in boron trifluoride diethyl etherate and trifluoroacetic acid. Journal of Electroanalytical Chemistry. 2008612,(2) , 191-200.