4-Nitroaniline, 98%
4-Nitroaniline, 98%
4-Nitroaniline, 98%
Thermo Scientific Chemicals
4-Nitroaniline, 98%
CAS: 100-01-6 | C6H6N2O2 | 138.13 g/mol
Have Questions?
Catalog NumberQuantity
ALFA10369.30250 g
Catalog number ALFA10369.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Nitroaniline
CAS100-01-6
Health Hazard 1H301+H311+H331-H373
Health Hazard 2GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c
View more
p-Nitroaniline is a a common chromophore and potentially useful syntetic reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
p-Nitroaniline is a a common chromophore and potentially useful syntetic reagent.
Solubility
Soluble in hot water and alcohol
Notes
Store at 4°C. Incompatible with oxidizing agents.
RUO – Research Use Only
E Kavitha; N Sundaraganesan and S Sebastian. Molecular structure, vibrational spectroscopic and HOMO, LUMO studies of 4-nitroaniline by density functional method.. Ind J Pure Appl Phys.2010, 48 (1), 20-30.