n-Pentadecane, 99%
n-Pentadecane, 99%
n-Pentadecane, 99%
Thermo Scientific Chemicals
n-Pentadecane, 99%
CAS: 629-62-9 | C15H32 | 212.42 g/mol
Have Questions?
Catalog NumberQuantity
ALFA10336.1850 g
Catalog number ALFA10336.18
Price (MYR)
439.00
EA
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
439.00
EA
Add to cart
Specifications
Chemical Name or Materialn-Pentadecane
CAS629-62-9
Health Hazard 1H304
Health Hazard 2GHS H Statement
H304
May be fatal if swallowed and enters airways.
Health Hazard 3P301+P310-P331-P501c
View more
n-pentadecane is used as a reference substance for gas chromatography. It is also used as standard materials for the determination of combustion quality of diesel. It is involved in the organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
n-pentadecane is used as a reference substance for gas chromatography. It is also used as standard materials for the determination of combustion quality of diesel. It is involved in the organic synthesis.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Vélez, C.; de Zárate, J. M. O.; Khayet, M. Thermal properties of n-pentadecane, n-heptadecane and n-nonadecane in the solid/liquid phase change region. Int. J. Therm. Sci. 2015, 94, 139-146.
  2. Fan, J.; Qi, M.; Fu, R.; Qu, L. Performance of graphene sheets as stationary phase for capillary gas chromatographic separations. J. Chromatogr. A 2015, 1399, 74-79.