Tri-n-butyl citrate, 99%
Tri-n-butyl citrate, 99%
Tri-n-butyl citrate, 99%
Tri-n-butyl citrate, 99%
Thermo Scientific Chemicals
Tri-n-butyl citrate, 99%
CAS: 77-94-1 | C18H32O7 | 360.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number ALFA10266.30
Price (MYR)
334.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
334.00
EA
Add to cart
Specifications
Chemical Name or MaterialTri-n-butyl citrate
CAS77-94-1
Melting Point-20°C
Recommended StorageAmbient temperatures
Density1.04
View more
Tri-n-butyl citrate is used in polishes, inks, nitrocellulose lacquers, as an anti-foam agent and as a plasticizer. It is also used as paint additives, coating additives and processing aids. In addition to this, it is added to the zein film to enhance its mechanical properties for industrial processing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tri-n-butyl citrate is used in polishes, inks, nitrocellulose lacquers, as an anti-foam agent and as a plasticizer. It is also used as paint additives, coating additives and processing aids. In addition to this, it is added to the zein film to enhance its mechanical properties for industrial processing.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Hufenus, R.; Reifler, F. A.; Fernández-Ronco, M. P.; Heuberger, M. Molecular orientation in melt-spun poly(3-hydroxybutyrate) fibers: Effect of additives, drawing and stress-annealing. Eur. Polym. J. 2015, 71, 12-26.
  2. Ivy, M. A.; Gallagher, L. T.; Ellington, A. D.; Anslyn, E. V. Exploration of plasticizer and plastic explosive detection and differentiation with serum albumin cross-reactive arrays. Chem. Sci. 2012, 3 (6), 1773-1779.