2,4-Octanedione, 98+%, Thermo Scientific™
2,4-Octanedione, 98+%, Thermo Scientific™
2,4-Octanedione, 98+%, Thermo Scientific™
2,4-Octanedione, 98+%, Thermo Scientific™
2,4-Octanedione, 98+%, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ALFA10161.0910 g
Catalog number ALFA10161.09
Price (MYR)
-
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or Material2,4-Octanedione
CAS14090-87-0
Recommended StorageAmbient temperatures
Density0.923
EINECS Number237-937-1
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.