5-Bromo-3-methoxysalicylaldehyde, 97%
5-Bromo-3-methoxysalicylaldehyde, 97%
5-Bromo-3-methoxysalicylaldehyde, 97%
5-Bromo-3-methoxysalicylaldehyde, 97%
Thermo Scientific Chemicals
5-Bromo-3-methoxysalicylaldehyde, 97%
CAS: 5034-74-2 | C8H7BrO3 | 231.045 g/mol
Have Questions?
Catalog NumberQuantity
ALFA10137.065 g
Catalog number ALFA10137.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material5-Bromo-3-methoxysalicylaldehyde
CAS5034-74-2
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c