Silicon(IV) oxide, 99.0% (metals basis)
Silicon(IV) oxide, 99.0% (metals basis)
Silicon(IV) oxide, 99.0% (metals basis)
Silicon(IV) oxide, 99.0% (metals basis)
Silicon(IV) oxide, 99.0% (metals basis)
Thermo Scientific Chemicals
Silicon(IV) oxide, 99.0% (metals basis)
CAS: 7631-86-9 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number ALF089709.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Boiling Point2230°C
Chemical Name or MaterialSilicon(IV) oxide
Melting Point1710°C
CAS7631-86-9
Recommended StorageAmbient temperatures
View more
Silicon(IV) oxide is used in the manufacture of glass for windows, drinking glasses, beverage bottles and many other materials. Optical fibers for telecommunication are made from silica. It is also used as a raw material for many ceramics such as earthenware, stoneware and porcelain. In addition, silica is a common additive in the production of foods and pharmaceutical products.