Niobium(IV) oxide, 99+% (metals basis)
Niobium(IV) oxide, 99+% (metals basis)
Niobium(IV) oxide, 99+% (metals basis)
Niobium(IV) oxide, 99+% (metals basis)
Thermo Scientific Chemicals
Niobium(IV) oxide, 99+% (metals basis)
CAS: 12034-59-2 | NbO2 | 124.90 g/mol
Have Questions?
Catalog NumberQuantity
ALF089692.0910 g
Catalog number ALF089692.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or MaterialNiobium(IV) oxide
CAS12034-59-2
Recommended StorageAmbient temperatures
Physical FormPowder
Quantity10 g
View more
Niobium(IV) oxide is used as a reducing agent, which reduces carbon dioxide and sulfur dioxide to carbon and sulfur respectively. It finds application in dielectric material for electronic capacitors. It is also used for manufacturing metal niobium metal, in optical glass and in electronic industry.