Phosphorous acid, 97%
Phosphorous acid, 97%
Phosphorous acid, 97%
Phosphorous acid, 97%
Thermo Scientific Chemicals
Phosphorous acid, 97%
CAS: 13598-36-2 | H3PO3
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
500 g
Catalog number ALF089407.A3
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialPhosphorous acid
CAS13598-36-2
Melting Point74°C
Boiling Point200°C (decomposition)
Health Hazard 1H290-H302-H314-H335
View more
Phosphorus acid is used as an intermediate in the preparation of other phosphorus compounds like potassium phosphite, ammonium phosphite and calcium phosphite. It is actively involved in the preparation of phosphites like aminotris(methylenephosphonic acid) (ATMP), 1-hydroxyethane 1,1-diphosphonic acid (HEDP) and 2-phosphonobutane-1,2,4-tricarboxylic Acid (PBTC), which find application in water treatment as a scale or corrosive inhibitor. It is also used in chemical reactions as a reducing agent. Its salt, lead phosphite is used as PVC stabilizer. It is also used as a precursor in the preparation of phosphine and as an intermediate in the preparation of other phosphorus compounds.