Ammonium sulfate, 99.95% (metals basis)
Ammonium sulfate, 99.95% (metals basis)
Ammonium sulfate, 99.95% (metals basis)
Ammonium sulfate, 99.95% (metals basis)
Thermo Scientific Chemicals
Ammonium sulfate, 99.95% (metals basis)
CAS: 7783-20-2 | H8N2O4S | 132.13 g/mol
Have Questions?
Catalog NumberQuantity
ALF089363.36500 g
Catalog number ALF089363.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium sulfate
Melting Point∽280°C (decomposition)
CAS7783-20-2
Recommended StorageAmbient temperatures
Physical FormCrystalline
View more
Ammonium sulfate is widely utilized as a fertilizer for alkaline soils. In biochemistry it is used for purification of proteins via ammonium sulfate precipitation method. As a food additive, it serves as an acidity regulator in flours and breads. It is used in the preparation of other ammonium salts, particularly ammonium persulfate. It is applied in several applications as a flame retardant and a wood preservative. It finds use as raw materials for various chemical products such as dyes and pharmaceutical products. A saturated solution of ammonium sulfate in heavy water is employed as an external standard in sulfur-33 NMR spectroscopy.