Amberlyst™ 15(H), ion exchange resin
Amberlyst™ 15(H), ion exchange resin
Amberlyst™ 15(H), ion exchange resin
Amberlyst™ 15(H), ion exchange resin
Thermo Scientific Chemicals
Amberlyst™ 15(H), ion exchange resin
Strongly acidic macroreticular resin | CAS: 39389-20-3 | C18H18O3S | 314.399 g/mol
Have Questions?
Catalog NumberQuantity
ALF089079.30250 g
Catalog number ALF089079.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmberlyst™ 15(H)
Physical FormPale brown beads
CAS39389-20-3
Recommended StorageAmbient temperatures
TSCAYes
View more
Amberlyst™ 15(H) is a strongly acidic cation exchange resin used as a heterogeneous acid catalysis and is suitable for non-aqueous catalysis. It is used to determine the mechanism of phenol alkylation using density functional theory (DFT). It is widely utilized as a column packing material for HPLC analysis. It is employed in separation media for Thin-layer chromatography (TLC) and paper chromatography. It is used in various chemical reactions like esterification, transesterification, Michael addition, Prins cyclization, Friedel-Crafts alkylation, acylation, cleavage of epoxides and crossed-aldol condensation.