Hydroxylamine sulfate, 99%
Hydroxylamine sulfate, 99%
Hydroxylamine sulfate, 99%
Hydroxylamine sulfate, 99%
Thermo Scientific Chemicals
Hydroxylamine sulfate, 99%
CAS: 10039-54-0 | H3NO·0.5H2SO4
Have Questions?
Catalog NumberQuantity
ALF088944.36500 g
Catalog number ALF088944.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialHydroxylamine sulfate
Melting Point∽170°C (decomposition)
CAS10039-54-0
Health Hazard 1H290-H302+H312-H315-H317-H319-H351-H373
Health Hazard 3P201-P202-P234-P260-P264b-P270-P272-P280-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P312-P330-P333+P313-P362-P390-P501c
View more
Hydroxylammonium sulfate is used to produce hydroxylamine-O-sulfonic acid from chlorosulfuric acid. It is widely used in organic synthesis for the conversion of aldehydes and ketones to oximes, carboxylic acids and their derivatives (e.g. esters) to hydroxamic acids, isocyanates to N-hydroxyureas and nitriles to amidoximes. It acts as a radical scavenger which is used to terminate the radical polymerization reactions. It finds application as an antioxidant in natural rubber. It is also used in photography as a stabilizer for color developers.