Starch indicator solution 1%, Acculute Standard Volumetric Solution, Final volume 1 L, Thermo Scientific™
Starch indicator solution 1%, Acculute Standard Volumetric Solution, Final volume 1 L, Thermo Scientific™
Starch indicator solution 1%, Acculute Standard Volumetric Solution, Final volume 1 L, Thermo Scientific™
Starch indicator solution 1%, Acculute Standard Volumetric Solution, Final volume 1 L, Thermo Scientific™
Thermo Scientific Chemicals
Starch indicator solution 1%, Acculute Standard Volumetric Solution, Final volume 1 L, Thermo Scientific™
For Iodometric Titrations
Have Questions?
Catalog number ALF088632.LQ
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS9005-84-9, 7732-18-5
Specifications
Chemical Name or MaterialStarch indicator solution
CAS9005-84-9
Physical FormLiquid,For iodometric titrations
CAS7732-18-5
Recommended StorageAmbient temperatures
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.