Bronze powder, -100 mesh, 99.0% (metals basis)
Bronze powder, -100 mesh, 99.0% (metals basis)
Thermo Scientific Chemicals
Bronze powder, -100 mesh, 99.0% (metals basis)
Have Questions?
Catalog NumberQuantity
ALF088359.A11 kg
Catalog number ALF088359.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialBronze
Health Hazard 1H228-H315-H319-H335
Health Hazard 3P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
AppearanceBronze colored powder
Recommended StorageAmbient temperatures
View more
Bronze powder suitable for a range of applications including resin-casting, decorative coatings and powder metallurgy. Also used for hydraulic seals, surface engineering, decorative, welding and brazing, diamond tooling.