Yttrium, AAS standard solution, Specpure™ Y 1000μg/mL
Yttrium, AAS standard solution, Specpure™ Y 1000μg/mL
Yttrium, AAS standard solution, Specpure™ Y 1000μg/mL
Yttrium, AAS standard solution, Specpure™ Y 1000μg/mL
Thermo Scientific Chemicals
Yttrium, AAS standard solution, Specpure™ Y 1000μg/mL
CAS: 7697-37-2 | HNO3 | 63.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALF088117.AP
Price (MYR)
543.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
543.00
EA
Add to cart
Specifications
Chemical Name or MaterialYttrium
CAS7697-37-2
Health Hazard 1H290-H314-H335
Health Hazard 3P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c