Rhodium, AAS standard solution, Specpure™ Rh 1000μg/mL
Rhodium, AAS standard solution, Specpure™ Rh 1000μg/mL
Thermo Scientific Chemicals
Rhodium, AAS standard solution, Specpure™ Rh 1000μg/mL
Have Questions?
Catalog NumberQuantity
ALF088090.AE100 mL
Catalog number ALF088090.AE
Price (MYR)
1,131.00
EA
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
1,131.00
EA
Add to cart
Specifications
Chemical Name or MaterialRhodium, AAS standard solution
Name NoteRh 1000μg/mL
Health Hazard 1H290-H314-H335
Health Hazard 2GHS H Statement
H314-H335
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Health Hazard 3P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c