Platinum, AAS standard solution, Specpure™ Pt 1000μg/mL
Platinum, AAS standard solution, Specpure™ Pt 1000μg/mL
Platinum, AAS standard solution, Specpure™ Pt 1000μg/mL
Platinum, AAS standard solution, Specpure™ Pt 1000μg/mL
Thermo Scientific Chemicals
Platinum, AAS standard solution, Specpure™ Pt 1000μg/mL
Form: Liquid | CAS: 7440-6-6 | Pt in 20% HCl
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALF088086.AP
Price (MYR)
1,979.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
1,979.00
EA
Add to cart
Specifications
Chemical Name or MaterialPlatinum
CAS7440-6-6
Health Hazard 1H290-H314-H335
Health Hazard 3P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c