Palladium, AAS standard solution, Specpure™ Pd 1000μg/mL, Thermo Scientific Chemicals
Palladium, AAS standard solution, Specpure™ Pd 1000μg/mL, Thermo Scientific Chemicals
Palladium, AAS standard solution, Specpure™ Pd 1000μg/mL, Thermo Scientific Chemicals
Palladium, AAS standard solution, Specpure™ Pd 1000μg/mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Palladium, AAS standard solution, Specpure™ Pd 1000μg/mL, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALF088085.AE100 mL
Catalog number ALF088085.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialPalladium, AAS standard solution
Name NotePd 1000μg/mL
Health Hazard 1H290-H314-H335
Health Hazard 2GHS H Statement
H314-H335
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Health Hazard 3P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c