Nickel, AAS standard solution, Specpure™ Ni 1000μg/mL
Nickel, AAS standard solution, Specpure™ Ni 1000μg/mL
Nickel, AAS standard solution, Specpure™ Ni 1000μg/mL
Nickel, AAS standard solution, Specpure™ Ni 1000μg/mL
Thermo Scientific Chemicals
Nickel, AAS standard solution, Specpure™ Ni 1000μg/mL
Have Questions?
Catalog NumberQuantity
ALF088082.AP500 mL
Catalog number ALF088082.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialNickel, AAS standard solution
Name NoteNi 1000μg/mL
Health Hazard 1H290-H314-H335-H350i-H360D
Health Hazard 2GHS H Statement
H314-H317-H208
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P234-P260-P264b-P271-P281-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c