Neodymium, AAS standard solution, Specpure™ Nd 1000μg/mL
Neodymium, AAS standard solution, Specpure™ Nd 1000μg/mL
Neodymium, AAS standard solution, Specpure™ Nd 1000μg/mL
Neodymium, AAS standard solution, Specpure™ Nd 1000μg/mL
Thermo Scientific Chemicals
Neodymium, AAS standard solution, Specpure™ Nd 1000μg/mL
CAS: 1313-97-9 | Nd2O3 | 336.48 g/mol
Have Questions?
Catalog NumberQuantity
ALF088081.AE100 mL
Catalog number ALF088081.AE
Price (MYR)
294.00
EA
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
294.00
EA
Add to cart
Specifications
Chemical Name or MaterialNeodymium
CAS1313-97-9
Health Hazard 1H290-H314-H335
CAS7697-37-2
Health Hazard 3P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c