Magnesium, AAS standard solution, Specpure™ Mg 1000μg/mL
Magnesium, AAS standard solution, Specpure™ Mg 1000μg/mL
Magnesium, AAS standard solution, Specpure™ Mg 1000μg/mL
Magnesium, AAS standard solution, Specpure™ Mg 1000μg/mL
Thermo Scientific Chemicals
Magnesium, AAS standard solution, Specpure™ Mg 1000μg/mL
CAS: 7439-95-4 | Mg | 24.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALF088077.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialMagnesium
CAS7439-95-4
Health Hazard 1H290-H314-H335
CAS7697-37-2
Health Hazard 3P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c