Gallium, AAS standard solution, Specpure™ Ga 1000μg/mL
Gallium, AAS standard solution, Specpure™ Ga 1000μg/mL
Thermo Scientific Chemicals
Gallium, AAS standard solution, Specpure™ Ga 1000μg/mL
Have Questions?
Catalog NumberQuantity
ALF088066.AP500 mL
Catalog number ALF088066.AP
Price (MYR)
702.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
702.00
EA
Add to cart
Specifications
Chemical Name or MaterialGallium, AAS standard solution
Name NoteGa 1000μg/mL
Health Hazard 1H290-H314-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c