Bismuth, AAS standard solution, Specpure™ Bi 1000μg/mL
Bismuth, AAS standard solution, Specpure™ Bi 1000μg/mL
Bismuth, AAS standard solution, Specpure™ Bi 1000μg/mL
Bismuth, AAS standard solution, Specpure™ Bi 1000μg/mL
Thermo Scientific Chemicals
Bismuth, AAS standard solution, Specpure™ Bi 1000μg/mL
CAS: 7440-69-9 | Bi | 208.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALF088054.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialBismuth
CAS7440-69-9
Health Hazard 1H290-H314-H335
CAS7732-18-5
Health Hazard 3P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c