Chlorosulfonic acid, typically 99%
Chlorosulfonic acid, typically 99%
Chlorosulfonic acid, typically 99%
Chlorosulfonic acid, typically 99%
Thermo Scientific Chemicals
Chlorosulfonic acid, typically 99%
CAS: 7790-94-5 | ClHO3S | 116.515 g/mol
Have Questions?
Catalog NumberQuantity
ALF087977.30250 g
Catalog number ALF087977.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
AppearanceYellow to brown liquid
Chemical Name or MaterialChlorosulfonic Acid
ColorYellow to Brown
CAS7790-94-5
Melting Point-80°C
View more
Chlorosulfonic acid is used as detergent and as an anti-contrail agent. It is used as an intermediate in the production of other substances such as pharmaceuticals and chemicals. It finds application in producing smoke screens.