Ammonium hydroxide, 25% NH{3}, 99.99% (metals basis)
Ammonium hydroxide, 25% NH{3}, 99.99% (metals basis)
Ammonium hydroxide, 25% NH{3}, 99.99% (metals basis)
Ammonium hydroxide, 25% NH{3}, 99.99% (metals basis)
Thermo Scientific Chemicals
Ammonium hydroxide, 25% NH{3}, 99.99% (metals basis)
CAS: 1336-21-6 | H5NO | 35.05 g/mol
Have Questions?
Catalog NumberQuantity
ALF087903.AK250 mL
Catalog number ALF087903.AK
Price (MYR)
975.00
EA
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (MYR)
975.00
EA
Add to cart
Specifications
Chemical Name or MaterialAmmonium hydroxide
ColorColorless
CAS1336-21-6
Health Hazard 1H314-H335
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more
Ammonium hydroxide is widely utilized as a leavening agent or acidity regulator in food production. It serves as a precursor to some alkyl amines and is also used in the tobacco industry for flavor enhancement and as a processing aid. During furniture making, it combines with tannic acid and is used to darken or stain wood by making it iron salts. In chemical laboratories, it used for qualitative inorganic analysis, as a complexant and as a base. It is used to clean gold, silver and platinum jewelry. It is an active component of Tollens' reagent (consisting of a solution of silver nitrate and ammonia) and is used to determine the presence of aldehyde or alpha-hydroxy ketone functional groups.