Nickel Aluminum, Raney∣r type non-activated
Nickel Aluminum, Raney∣r type non-activated
Nickel Aluminum, Raney∣r type non-activated
Nickel Aluminum, Raney∣r type non-activated
Nickel Aluminum, Raney∣r type non-activated
CAS: 12635-29-9 | AlNi | 85.675 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
50 g
Catalog number ALF087676.36
Price (MYR)
-
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialNickel Aluminum
ColorBlack to Gray
Name NoteRaney™ type non-activated
CAS12635-29-9
Health Hazard 1H261-H317-H334-H350-H372
View more
Nickel Aluminum is used for the manufacture of aluminum-nickel alloy powder, and it is used in the chemical, pharmaceutical.