Bromine liquid, 99.99+% (metals basis)
Bromine liquid, 99.99+% (metals basis)
Bromine liquid, 99.99+% (metals basis)
Bromine liquid, 99.99+% (metals basis)
Thermo Scientific Chemicals
Bromine liquid, 99.99+% (metals basis)
CAS: 7726-95-6 | Br | 79.91 g/mol
Have Questions?
Catalog NumberQuantity
ALF087614.AK250 mL
Catalog number ALF087614.AK
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialBromine liquid
CAS7726-95-6
ColorRed to Brown
Health Hazard 1H314-H330
Health Hazard 3P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c