Sodium bromide, 99.99% (metals basis)
Sodium bromide, 99.99% (metals basis)
Sodium bromide, 99.99% (metals basis)
Sodium bromide, 99.99% (metals basis)
Thermo Scientific Chemicals
Sodium bromide, 99.99% (metals basis)
CAS: 7647-15-6 | BrNa | 102.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALF087604.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Boiling Point1390°C
Chemical Name or MaterialSodium bromide
Melting Point755°C
CAS7647-15-6
Recommended StorageAmbient temperatures
View more
Sodium bromide is used in pharmaceutical applications, used in photographic processing and chemical intermediate for production of chemicals and bromides. It is widely used as an anticonvulsant. Also used for the preparation of drilling fluids, completion fluids and drilling muds formulations.