Phenylmagnesium bromide, 3M in ether
Phenylmagnesium bromide, 3M in ether
Phenylmagnesium bromide, 3M in ether
Phenylmagnesium bromide, 3M in ether
Thermo Scientific Chemicals
Phenylmagnesium bromide, 3M in ether
Grignard reagent | CAS: 100-58-3 | C6H5BrMg | 181.32 g/mol
Have Questions?
Catalog NumberQuantity
ALF087326.AP500 mL
Catalog number ALF087326.AP
Price (MYR)
1,254.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
1,254.00
EA
Add to cart
Specifications
Chemical Name or MaterialPhenylmagnesium bromide
CAS100-58-3
Health Hazard 1H224-H314-H335-H336
Health Hazard 2GHS H Statement
H224-H261-H314-H336
Extremely flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
View more
Grignard reagent Phenylmagnesium bromide is an important reagent called as Grignard reagent used in the synthesis of organic and oraganometallic reactions. It is used to prepare phenyl aryl sulphides, sulphoxides, and sulphones. It acts as a strong acid and Lewis acid and undergoes addition reaction with many unsaturated functional groups. It is involved in the synthesis of end-functionalized regioregular poly(3-alkylthiophene)s, 1,3,3-trimethyl-6-phenyl-2-oxabicyclo[2.2.2]octan-6-ol, o-substituted benzophenones and 6-benzyl-1,3,3-trimethyl-2-oxabicyclo[2.2.2]octan-6-ol. In addition to this, it is used for the monalkylation of primary amines to get secondary amines.