Dimethyltin dichloride
Dimethyltin dichloride
Dimethyltin dichloride
Dimethyltin dichloride
Thermo Scientific Chemicals
Dimethyltin dichloride
CAS: 753-73-1 | C2H6Cl2Sn | 219.68 g/mol
Have Questions?
Catalog NumberQuantity
ALF071142.1850 g
Catalog number ALF071142.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDimethyltin dichloride
CAS753-73-1
Health Hazard 1H301+H311-H314-H330-H360-H372
Health Hazard 2GHS H Statement
H300-H310-H330-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P201-P202-P260-P264b-P270-P271-P281-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
View more
Used as a pharmaceutical intermediate. It is employed as a coating, plastic and rubber auxilliary agent. Used as methyl Tin stabilizer intermediates, catalysts for esterification reaction, raw material for glass coating.