Starch indicator solution 1%, w/v aqueous solution (for iodometric titrations)
Starch indicator solution 1%, w/v aqueous solution (for iodometric titrations)
Starch indicator solution 1%, w/v aqueous solution (for iodometric titrations)
Starch indicator solution 1%, w/v aqueous solution (for iodometric titrations)
Thermo Scientific Chemicals
Starch indicator solution 1%, w/v aqueous solution (for iodometric titrations)
For iodometric titrations | CAS: 9005-84-9 | C12H22O11 | 342.297 g/mol
Have Questions?
Catalog NumberQuantity
ALF047209.AP500 mL
Catalog number ALF047209.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialStarch indicator solution 1%
Name Notew/v aqueous solution for iodometric titrations
CAS9005-84-9
Physical FormLiquid for iodometric titrations