Scandium(III) bromide, ultra dry, 99.99% (metals basis)
Scandium(III) bromide, ultra dry, 99.99% (metals basis)
Scandium(III) bromide, ultra dry, 99.99% (metals basis)
Scandium(III) bromide, ultra dry, 99.99% (metals basis)
Thermo Scientific Chemicals
Scandium(III) bromide, ultra dry, 99.99% (metals basis)
CAS: 13465-59-3 | Br3Sc | 284.67 g/mol
Have Questions?
Catalog NumberQuantity
ALF047199.031 g
Catalog number ALF047199.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialScandium(III) bromide
Name NoteUltra dry
CAS13465-59-3
Storage Note 1
  • Ampouled under argon
Recommended StorageAmbient temperatures
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only