Graphene nanoplatelets aggregates, sub-micronparticles, S.A. 500 m^2/g
Graphene nanoplatelets aggregates, sub-micronparticles, S.A. 500 m^2/g
Graphene nanoplatelets aggregates, sub-micronparticles, S.A. 500 m^2/g
Graphene nanoplatelets aggregates, sub-micronparticles, S.A. 500 m^2/g
Graphene nanoplatelets aggregates, sub-micronparticles, S.A. 500 m^2/g
CAS: 7782-42-5 | C | 12.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALF047132.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialGraphene nanoplatelets aggregates
Name Notesub-micronparticles
CAS7782-42-5
Health Hazard 1H319-H335
Health Hazard 2GHS H Statement
H319-H335