Silicon(IV) oxide, 99.95% (metals basis)
Silicon(IV) oxide, 99.95% (metals basis)
Silicon(IV) oxide, 99.95% (metals basis)
Silicon(IV) oxide, 99.95% (metals basis)
Thermo Scientific Chemicals
Silicon(IV) oxide, 99.95% (metals basis)
CAS: 7631-86-9 | O2Si | 60.08 g/mol
Have Questions?
Catalog NumberQuantity
ALF047098.1425 g
Catalog number ALF047098.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Boiling Point2230°C
Chemical Name or MaterialSilicon (IV) oxide
Melting Point1710°C
CAS7631-86-9
ColorWhite
View more
Used as sodium stabilizing counterion, for preparation of silica catalytic supports. Also used in glass manufacturing, foundry sand casting, metallurgical flux sand, ceramics sand, gas recovery, paint and coatings, filtration sand, chromatographic column treatment, wood treatment, inks, desiccants and powder metal molding. Its is principle gateway in electronic industries.