Viscosity standard, Specpure™, nominally 50cSt @ 40 and 7.3cSt @ 100
Viscosity standard, Specpure™, nominally 50cSt @ 40 and 7.3cSt @ 100
Thermo Scientific Chemicals
Viscosity standard, Specpure™, nominally 50cSt @ 40 and 7.3cSt @ 100
CAS: 8042-47-5 | C16H10N2Na2O7S2 | 452.363 g/mol
Have Questions?
Catalog NumberQuantity
ALF047013.AP500 mL
Catalog number ALF047013.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialViscosity standard
Name NoteNominally 50cSt at 40 and 7.3cSt at 100
CAS8042-47-5
Health Hazard 1H304
Health Hazard 3P301+P310-P331-P501c