Cobalt nanopowder, APS 25-30nm, 99.8% (metals basis)
Cobalt nanopowder, APS 25-30nm, 99.8% (metals basis)
Cobalt nanopowder, APS 25-30nm, 99.8% (metals basis)
Cobalt nanopowder, APS 25-30nm, 99.8% (metals basis)
Thermo Scientific Chemicals
Cobalt nanopowder, APS 25-30nm, 99.8% (metals basis)
CAS: 7440-48-4 | Co | 58.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALF046347.14
Price (MYR)
2,225.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
2,225.00
EA
Add to cart
Specifications
Chemical Name or MaterialCobalt nanopowder
CAS7440-48-4
Health Hazard 1H228-H317-H334-H351
Health Hazard 2GHS H Statement
H228-H334-H351-H317
Flammable solid.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing cancer.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P210-P240-P241-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P370+P378q-P501c