Ammonium pentaborate tetrahydrate, 98%
Ammonium pentaborate tetrahydrate, 98%
Ammonium pentaborate tetrahydrate, 98%
Ammonium pentaborate tetrahydrate, 98%
Thermo Scientific Chemicals
Ammonium pentaborate tetrahydrate, 98%
CAS: 12046-04-7 | B5H44N9O19-6 | 528.446 g/mol
Have Questions?
Catalog NumberQuantity
ALF046323.36500 g
Catalog number ALF046323.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium pentaborate tetrahydrate
Melting Point110°C
CAS12046-04-7
ColorWhite
Health Hazard 1H361
View more
Ammonium pentaborate is used in the preparation of both wet and dry electrolytic capacitors, flame proofing, gunning and patching compounds for extending the life of basic refractories. Used in the preparation of special welding, soldering and brazing fluxes for stainless steel or various non-ferrous metals, where alkali borates cannot be used.