Fullerene powder, 99% C{6}{0}
Fullerene powder, 99% C{6}{0}
Fullerene powder, 99% C{6}{0}
Fullerene powder, 99% C{6}{0}
Fullerene powder, 99% C{6}{0}
Thermo Scientific Chemicals
Fullerene powder, 99% C{6}{0}
CAS: 99685-96-8 | C60 | 720.66 g/mol
Have Questions?
Catalog NumberQuantity
ALF046311.065 g
Catalog number ALF046311.06
Price (MYR)
7,795.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
7,795.00
EA
Add to cart
Specifications
Chemical Name or MaterialFullerene powder
CAS99685-96-8
Health Hazard 1H319-H335
Health Hazard 2GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
View more
It is used in optical computing, optical memories, optical signal processing and controlling applications respects, C60 and its derivatives can be widely used in photovoltaic applications, magnetic resonance imaging, anti-HIV drugs, anti-cancer drugs, chemotherapy drugs, cosmetic additives, research and other areas.