Antimony(III) telluride, 99.999% (metals basis)
Antimony(III) telluride, 99.999% (metals basis)
Antimony(III) telluride, 99.999% (metals basis)
Antimony(III) telluride, 99.999% (metals basis)
Thermo Scientific Chemicals
Antimony(III) telluride, 99.999% (metals basis)
CAS: 1327-50-0 | Sb2Te3 | 626.32 g/mol
Have Questions?
Catalog NumberQuantity
ALF045922.0910 g
Catalog number ALF045922.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAntimony (III) telluride
Melting Point629°C
CAS1327-50-0
ColorGray
Health Hazard 1H302+H332
View more
Antimony(III) telluride is used as both n-type and p-type semiconductors in association with appropriate dopant. It reacts with germanium telluride and forms the pseudobinary intermetallic system. It is used in solid state refrigerators due to its large thermoelectric effect. Further, it is an active component of thermoelectric materials.