Ytterbium powder, -200 mesh, 99.9% (REO)
Ytterbium powder, -200 mesh, 99.9% (REO)
Ytterbium powder, -200 mesh, 99.9% (REO)
Ytterbium powder, -200 mesh, 99.9% (REO)
Thermo Scientific Chemicals
Ytterbium powder, -200 mesh, 99.9% (REO)
CAS: 7440-64-4 | Yb | 173.05 g/mol
Have Questions?
Catalog NumberQuantity
ALF045920.0910 g
Catalog number ALF045920.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or MaterialYtterbium powder
ColorSilver
CAS7440-64-4
Health Hazard 1H228-H260
Health Hazard 2GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under Argon
RUO – Research Use Only